missing translation for 'onlineSavingsMsg'
Learn More

2-Bromohexanoic acid, 97%

Catalog No. AAB2422822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2422822 100 g
AAB2422814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2422822 Supplier Thermo Scientific Chemicals Supplier No. B2422822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 616-05-7 | C6H11BrO2 | 195.056 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 616-05-7
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.056
MDL Number MFCD00004218
InChI Key HZTPKMIMXLTOSK-UHFFFAOYSA-N
Synonym 2-bromocaproic acid, dl-2-bromohexanoic acid, hexanoic acid, 2-bromo, alpha-bromohexanoic acid, alpha-bromo-n-caproic acid, .alpha.-bromo-n-caproic acid, 2-bromo-hexanoic acid, .alpha.-bromocaproic acid, .alpha.-bromohexanoic acid, 2-bromo-n-hexanoic acid
PubChem CID 12013
IUPAC Name 2-bromohexanoic acid
SMILES CCCCC(C(=O)O)Br

Specifications

Melting Point 3°C to 5°C
Density 1.37
Boiling Point 136°C to 138°C (18 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.472
Quantity 100 g
UN Number UN3265
Merck Index 14,1415
Formula Weight 195.06
Percent Purity 97%
Chemical Name or Material 2-Bromohexanoic acid
Show More Show Less
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 210-461-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only