missing translation for 'onlineSavingsMsg'
Learn More

2-Bromohexane (contains 3-Bromohexane) (stabilized with Copper chip) 70.0+%, TCI America™

Catalog No. B123525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B123525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B123525G Supplier TCI America Supplier No. B123525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3377-86-4
Molecular Formula C6H13Br
Molecular Weight (g/mol) 165.074
MDL Number MFCD00039183
InChI Key NEBYCXAKZCQWAW-UHFFFAOYSA-N
Synonym hexane, 2-bromo, 2-hexyl bromide, dl, 2-bromo-hexane, acmc-209t5g, nebycxakzcqwaw-uhfffaoysa
PubChem CID 18805
IUPAC Name 2-bromohexane
SMILES CCCCC(C)Br

Specifications

Color Yellow
Quantity 25 g
UN Number 1993
Formula Weight 165.07
Percent Purity ≥70.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromohexane (contains 3-Bromohexane) (stabilized with Copper chip)
EINECSNumber (2)-0067
TSCA Yes