missing translation for 'onlineSavingsMsg'
Learn More

2-Bromofluorobenzene 99.0+%, TCI America™

Catalog No. B0883500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0883500G 500 g
B088325G 25 g
B0883100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0883500G Supplier TCI America Supplier No. B0883500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1072-85-1
Molecular Formula C6H4BrF
Molecular Weight (g/mol) 175
MDL Number MFCD00000282
InChI Key IPWBFGUBXWMIPR-UHFFFAOYSA-N
Synonym 2-bromofluorobenzene, o-bromofluorobenzene, 2-fluorobromobenzene, o-fluorobromobenzene, 2-bromfluorbenzen, benzene, 1-bromo-2-fluoro, 1-fluoro-2-bromobenzene, benzene, 1-fluoro-2-bromo, ofbb, benzene, bromofluoro
PubChem CID 61259
IUPAC Name 1-bromo-2-fluorobenzene
SMILES C1=CC=C(C(=C1)F)Br

Specifications

Melting Point -8°C
Color Yellow
Boiling Point 158°C
Quantity 500 g
UN Number 1993
Formula Weight 175.00
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromofluorobenzene
RTECSNumber DA1050000
TSCA Yes