missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromofluorene 98.0+%, TCI America™
$76.43 - $488.38
Chemical Identifiers
| CAS | 1133-80-8 |
|---|---|
| Molecular Formula | C13H9Br |
| Molecular Weight (g/mol) | 245.119 |
| MDL Number | MFCD00001115 |
| InChI Key | FXSCJZNMWILAJO-UHFFFAOYSA-N |
| Synonym | 2-bromofluorene, 9h-fluorene, 2-bromo, 2-brom-9h-fluoren, 2-bromofluorene, tech., #, acmc-1c7b2, ksc181a7p, 2-bromofluorene 5g, 4-bromo-2,2'-methylenebiphenyl |
| PubChem CID | 14336 |
| IUPAC Name | 2-bromo-9H-fluorene |
| SMILES | C1C2=CC=CC=C2C3=C1C=C(C=C3)Br |
Chemical Identifiers
| 1133-80-8 | |
| 245.119 | |
| FXSCJZNMWILAJO-UHFFFAOYSA-N | |
| 14336 | |
| C1C2=CC=CC=C2C3=C1C=C(C=C3)Br |
| C13H9Br | |
| MFCD00001115 | |
| 2-bromofluorene, 9h-fluorene, 2-bromo, 2-brom-9h-fluoren, 2-bromofluorene, tech., #, acmc-1c7b2, ksc181a7p, 2-bromofluorene 5g, 4-bromo-2,2'-methylenebiphenyl | |
| 2-bromo-9H-fluorene |
Specifications
| 1133-80-8 | |
| White-Yellow | |
| C13H9Br | |
| 5 g | |
| FXSCJZNMWILAJO-UHFFFAOYSA-N | |
| 2-bromo-9H-fluorene | |
| 14336 | |
| ≥98.0% (GC) | |
| 2-Bromofluorene |
| 114°C | |
| 185°C | |
| MFCD00001115 | |
| 2-bromofluorene, 9h-fluorene, 2-bromo, 2-brom-9h-fluoren, 2-bromofluorene, tech., #, acmc-1c7b2, ksc181a7p, 2-bromofluorene 5g, 4-bromo-2,2'-methylenebiphenyl | |
| C1C2=CC=CC=C2C3=C1C=C(C=C3)Br | |
| 245.119 | |
| 245.12 | |
| Crystalline Powder |
Safety and Handling
TSCA : No