missing translation for 'onlineSavingsMsg'
Learn More

2-Bromofluorene 98.0+%, TCI America™

Catalog No. B30695G
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30695G 5 g
B306925G 25 g
B3069100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B30695G Supplier TCI America Supplier No. B30695G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1133-80-8
Molecular Formula C13H9Br
Molecular Weight (g/mol) 245.119
MDL Number MFCD00001115
InChI Key FXSCJZNMWILAJO-UHFFFAOYSA-N
Synonym 2-bromofluorene, 9h-fluorene, 2-bromo, 2-brom-9h-fluoren, 2-bromofluorene, tech., #, acmc-1c7b2, ksc181a7p, 2-bromofluorene 5g, 4-bromo-2,2'-methylenebiphenyl
PubChem CID 14336
IUPAC Name 2-bromo-9H-fluorene
SMILES C1C2=CC=CC=C2C3=C1C=C(C=C3)Br

Specifications

Melting Point 114°C
Color White-Yellow
Boiling Point 185°C
Quantity 5 g
Formula Weight 245.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromofluorene