missing translation for 'onlineSavingsMsg'
Learn More

2-Bromofluorene, 95%

Catalog No. AAL0758906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0758906 5 g
AAL0758914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0758906 Supplier Thermo Scientific Chemicals Supplier No. L0758906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1133-80-8 | C13H9Br | 245.119 g/mol

2-Bromofluorene acts as a nonintrusive end-capping reagent in order to control molecular weights as well as in the generation of well-defined oligomers. It is used to prepare 2-bromo-9-cyclohepta-2,4,6-trienyl-9H-fluorene and 9,9-dihexylflorene. In addition to this, it is used in the preparation of poly(di-n-hexylfluorene)s end capped with 2-bromofluorene, 2-bromo-9,9-di-n-hexylfluorene and 9-bromoanthracene through Ni(0) mediated polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromofluorene acts as a nonintrusive end-capping reagent in order to control molecular weights as well as in the generation of well-defined oligomers. It is used to prepare 2-bromo-9-cyclohepta-2,4,6-trienyl-9H-fluorene and 9,9-dihexylflorene. In addition to this, it is used in the preparation of poly(di-n-hexylfluorene)s end capped with 2-bromofluorene, 2-bromo-9,9-di-n-hexylfluorene and 9-bromoanthracene through Ni(0) mediated polymerization.

Solubility
Soluble in chloroform and ethyl acetate.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1133-80-8
Molecular Formula C13H9Br
Molecular Weight (g/mol) 245.119
MDL Number MFCD00001115
InChI Key FXSCJZNMWILAJO-UHFFFAOYSA-N
Synonym 2-bromofluorene, 9h-fluorene, 2-bromo, 2-brom-9h-fluoren, 2-bromofluorene, tech., #, acmc-1c7b2, ksc181a7p, 2-bromofluorene 5g, 4-bromo-2,2'-methylenebiphenyl
PubChem CID 14336
IUPAC Name 2-bromo-9H-fluorene
SMILES C1C2=CC=CC=C2C3=C1C=C(C=C3)Br

Specifications

Melting Point 110°C to 114°C
Boiling Point 185°C (135 mmHg)
Quantity 5 g
Beilstein 1946701
Solubility Information Soluble in chloroform and ethyl acetate.
Formula Weight 245.12
Percent Purity 95%
Chemical Name or Material 2-Bromofluorene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-480-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only