missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromoethylamine Hydrobromide 98.0+%, TCI America™
Supplier: TCI America B0591100G
Specifications
| 2-Bromoethylamine Hydrobromide | |
| 175°C | |
| MFCD00012886 | |
| 2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromide | |
| C(CBr)N.Br | |
| 204.893 | |
| 204.89 | |
| Crystalline Powder |
| 2576-47-8 | |
| C2H7Br2N | |
| 100 g | |
| WJAXXWSZNSFVNG-UHFFFAOYSA-N | |
| 2-bromoethanamine;hydrobromide | |
| 2774217 | |
| ≥98.0% (N,T) |
Chemical Identifiers
| 2576-47-8 | |
| 204.893 | |
| WJAXXWSZNSFVNG-UHFFFAOYSA-N | |
| 2774217 | |
| C(CBr)N.Br |
| C2H7Br2N | |
| MFCD00012886 | |
| 2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromide | |
| 2-bromoethanamine;hydrobromide |
Safety and Handling
RTECSNumber : KQ8225000
TSCA : Yes