missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethylamine Hydrobromide 98.0+%, TCI America™

Catalog No. B059125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B059125G 25 g
B0591100G 100 g
B0591500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B059125G Supplier TCI America Supplier No. B059125G

Chemical Identifiers

CAS 2576-47-8
Molecular Formula C2H7Br2N
Molecular Weight (g/mol) 204.893
MDL Number MFCD00012886
InChI Key WJAXXWSZNSFVNG-UHFFFAOYSA-N
Synonym 2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromide
PubChem CID 2774217
IUPAC Name 2-bromoethanamine;hydrobromide
SMILES C(CBr)N.Br

Specifications

Melting Point 175°C
Quantity 25 g
Formula Weight 204.89
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoethylamine Hydrobromide
RTECSNumber KQ8225000
TSCA Yes