Learn More
2-Bromoethylamine hydrobromide, 98+%
CAS: 2576-47-8 | C2H7Br2N | 204.893 g/mol
Supplier: Thermo Scientific Chemicals A176250B
Description
2-Bromoethylamine hydrobromide is used to construct C2-symmetric imidazolidinylidene ligands with a dioxolane backbone. It acts as a building block which is used for proteomics research. It is also used as organic and pharmaceutical intermediates for the preparation of first-aid medicines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromoethylamine hydrobromide is used to construct C2-symmetric imidazolidinylidene ligands with a dioxolane backbone. It acts as a building block which is used for proteomics research. It is also used as organic and pharmaceutical intermediates for the preparation of first-aid medicines.
Solubility
Soluble in water.
Notes
Hygroscopic and moisture sensitive. Incompatible with strong oxidizing agents.
Specifications
| 2-Bromoethylamine hydrobromide | |
| 172°C to 176°C | |
| MFCD00012886 | |
| 3607605 | |
| 2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromide | |
| WJAXXWSZNSFVNG-UHFFFAOYSA-N | |
| 2-bromoethanamine;hydrobromide | |
| 2774217 | |
| ≥98% |
| 2576-47-8 | |
| C2H7Br2N | |
| 1000 g | |
| Hygroscopic | |
| Soluble in water. | |
| C(CBr)N.Br | |
| 204.893 | |
| 204.9 |
Chemical Identifiers
| 2576-47-8 | |
| 204.893 | |
| WJAXXWSZNSFVNG-UHFFFAOYSA-N | |
| 2774217 | |
| C(CBr)N.Br |
| C2H7Br2N | |
| MFCD00012886 | |
| 2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromide | |
| 2-bromoethanamine;hydrobromide |
Safety and Handling
GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
H302-H315-H317-H319-H335
EINECSNumber : 219-924-2
RTECSNumber : KQ8225000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only