missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethyl Benzoate 98.0+%, TCI America™

Catalog No. B006525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B006525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B006525ML Supplier TCI America Supplier No. B006525ML
Only null left

Chemical Identifiers

CAS 939-54-8
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00059462
InChI Key KNBBDZULQFKSIE-UHFFFAOYSA-N
Synonym Benzoic Acid 2-Bromoethyl Ester
PubChem CID 523090
IUPAC Name 2-bromoethyl benzoate
SMILES BrCCOC(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 285°C
Quantity 25 mL
Formula Weight 229.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromoethyl Benzoate