missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromoethyl)benzene 98.0+%, TCI America™

Catalog No. p-7118551
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
B059425G 25 g
B0594100G 100 g
B0594500G 500 g
3 options

Catalog No. B059425G

Supplier: TCI America B059425G

Only null left

Chemical Identifiers

CAS 103-63-9
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000240
InChI Key WMPPDTMATNBGJN-UHFFFAOYSA-N
Synonym 2-bromoethyl benzene, phenethyl bromide, 2-phenylethyl bromide, 1-bromo-2-phenylethane, phenylethyl bromide, benzene, 2-bromoethyl, 2-phenethyl bromide, beta-bromoethylbenzene, 2-phenyl-1-bromoethane, phenethylbromide
PubChem CID 7666
IUPAC Name 2-bromoethylbenzene
SMILES C1=CC=C(C=C1)CCBr

Specifications

Color Yellow
Boiling Point 220°C
Quantity 25 g
Formula Weight 185.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (2-Bromoethyl)benzene
EINECSNumber (3)-0035
RTECSNumber CY9032000
TSCA Yes
Recommended Storage Refrigerator