missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromoethyl)benzene 98.0+%, TCI America™

Catalog No. B0594500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0594500G 500 g
B059425G 25 g
B0594100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0594500G Supplier TCI America Supplier No. B0594500G
Only null left

Chemical Identifiers

CAS 103-63-9
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000240
InChI Key WMPPDTMATNBGJN-UHFFFAOYSA-N
Synonym 2-bromoethyl benzene, phenethyl bromide, 2-phenylethyl bromide, 1-bromo-2-phenylethane, phenylethyl bromide, benzene, 2-bromoethyl, 2-phenethyl bromide, beta-bromoethylbenzene, 2-phenyl-1-bromoethane, phenethylbromide
PubChem CID 7666
IUPAC Name 2-bromoethylbenzene
SMILES C1=CC=C(C=C1)CCBr

Specifications

Color Yellow
Boiling Point 220°C
Quantity 500 g
Formula Weight 185.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (2-Bromoethyl)benzene
EINECSNumber (3)-0035
RTECSNumber CY9032000
TSCA Yes
Recommended Storage Refrigerator