missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethyl Acetate 98.0+%, TCI America™

Catalog No. A0023500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0023500G 500 g
A002325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0023500G Supplier TCI America Supplier No. A0023500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 927-68-4
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00000235
InChI Key RGHQKFQZGLKBCF-UHFFFAOYSA-N
Synonym bromoethyl acetate, 1-acetoxy-2-bromoethane, 2-bromoethylacetate, ethanol, 2-bromo-, acetate, 1-bromo-2-acetoxyethane, acetic acid 2-bromoethyl ester, unii-nrz256o7qb, ethanol, 2-bromo-, 1-acetate, beta-bromoethyl acetate, nrz256o7qb
PubChem CID 13564
IUPAC Name 2-bromoethyl acetate
SMILES CC(=O)OCCBr

Specifications

Color Yellow
Boiling Point 162°C
Quantity 500 g
Formula Weight 167.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromoethyl Acetate
RTECSNumber KJ8227000
TSCA Yes