missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethanol, MilliporeSigma™ Supelco™

Catalog No. 111020492
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020492 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020492 Supplier Merck Emd Millipore Supplier No. 442334
Only null left

2-Bromoethanol is one of the degradation by-products of commonly used preservative, bronopol.

Featured Industries: Environmental

Specifications

Packaging Ampule
CAS 540-51-2
Vapor Pressure 2.4 mmHg (20°C)
Vapor Density 4.3 (vs. air)
Density 1.763 g/mL (at 25°C)
Flash Point 110°C
Physical Form Neat
Linear Formula BrCH2 CH2 OH
MDL Number MFCD00002827
Boiling Point 56°C to 57°C (20 mmHg)
Synonym Ethylene bromohydrin
Type Organic Pollutant Standard
Quantity 1000 mg
Formula Weight 124.96
Grade Analytical Standard
Show More Show Less