missing translation for 'onlineSavingsMsg'
Learn More

2-Bromodibenzofuran 98.0+%, TCI America™

Catalog No. B44595G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44595G 5 g
B44591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44595G Supplier TCI America Supplier No. B44595G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 86-76-0
Molecular Formula C12H7BrO
Molecular Weight (g/mol) 247.091
MDL Number MFCD00092338
InChI Key CRJISNQTZDMKQD-UHFFFAOYSA-N
Synonym 2-bromodibenzo b,d furan, 2-bromo-dibenzofuran, dibenzofuran, 2-bromo, dibenzofuran, 2-bromo-8ci 9ci, 2-bromodibenzo b furan, 4-bromo-8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0 2 , trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0?,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0^ 2,7 trideca-1 13 ,2,4,6,9,11-hexaene
PubChem CID 6856
IUPAC Name 2-bromodibenzofuran
SMILES C1=CC=C2C(=C1)C3=C(O2)C=CC(=C3)Br

Specifications

Melting Point 112°C
Color White
Boiling Point 220°C
Quantity 5 g
Formula Weight 247.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromodibenzofuran
EINECSNumber (5)-0022
TSCA No