accessibility menu, dialog, popup

UserName

BrO

CAS RN 86-76-0

IUPAC Name: 4-bromo-8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Synonyms: 2-bromodibenzofuran
Molecular Weight (g/mol): 247.09
Molecular Formula: C12H7BrO
InChi Key: CRJISNQTZDMKQD-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(OC3=C2C=CC=C3)C=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)