missing translation for 'onlineSavingsMsg'
Learn More

2-Bromodibenzofuran, 98%, Thermo Scientific Chemicals

$287.74 - $287.74

Chemical Identifiers

CAS 86-76-0
Molecular Formula C12H7BrO
Molecular Weight (g/mol) 247.091
MDL Number MFCD00092338
InChI Key CRJISNQTZDMKQD-UHFFFAOYSA-N
Synonym 2-bromodibenzo b,d furan, 2-bromo-dibenzofuran, dibenzofuran, 2-bromo, dibenzofuran, 2-bromo-8ci 9ci, 2-bromodibenzo b furan, 4-bromo-8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0 2 , trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0?,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0^ 2,7 trideca-1 13 ,2,4,6,9,11-hexaene
PubChem CID 6856
IUPAC Name 2-bromodibenzofuran
SMILES C1=CC=C2C(=C1)C3=C(O2)C=CC(=C3)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6459603
View Documents
Thermo Scientific Chemicals
H6459603
1 g
Each for $287.74
Only null left
 
Description

Description

2-Bromodibenzofuran is used as a reagent in Friedel-Crafts acylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromodibenzofuran is used as a reagent in Friedel-Crafts acylation reactions.

Solubility
Soluble in hot methanol.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

86-76-0
247.091
CRJISNQTZDMKQD-UHFFFAOYSA-N
6856
C1=CC=C2C(=C1)C3=C(O2)C=CC(=C3)Br
C12H7BrO
MFCD00092338
2-bromodibenzo b,d furan, 2-bromo-dibenzofuran, dibenzofuran, 2-bromo, dibenzofuran, 2-bromo-8ci 9ci, 2-bromodibenzo b furan, 4-bromo-8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0 2 , trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0?,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0^ 2,7 trideca-1 13 ,2,4,6,9,11-hexaene
2-bromodibenzofuran

Specifications

86-76-0
C12H7BrO
1 g
2-bromodibenzo b,d furan, 2-bromo-dibenzofuran, dibenzofuran, 2-bromo, dibenzofuran, 2-bromo-8ci 9ci, 2-bromodibenzo b furan, 4-bromo-8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0 2 , trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0?,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0^ 2,7 trideca-1 13 ,2,4,6,9,11-hexaene
CRJISNQTZDMKQD-UHFFFAOYSA-N
2-bromodibenzofuran
6856
98%
2-Bromodibenzofuran
108°C to 111°C
MFCD00092338
142199
Soluble in hot methanol.
C1=CC=C2C(=C1)C3=C(O2)C=CC(=C3)Br
247.091
247.09
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only