missing translation for 'onlineSavingsMsg'
Learn More

2-Bromocinnamic Acid 98.0+%, TCI America™

Catalog No. p-7119371
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19435G 5 g
B194325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19435G Supplier TCI America Supplier No. B19435G

Chemical Identifiers

CAS 7345-79-1
Molecular Formula C9H7BrO2
Molecular Weight (g/mol) 227.06
MDL Number MFCD00016836
InChI Key OMHDOOAFLCMRFX-UHFFFAOYSA-N
Synonym 2-bromocinnamic acid, trans-2-bromocinnamic acid, o-bromocinnamic acid, 3-2-bromophenyl acrylic acid, bromocinnamic acid,2, 2e-3-2-bromophenyl prop-2-enoic acid, e-3-2-bromophenyl acrylic acid, 3-2-bromo-phenyl-acrylic acid, e-3-2-bromophenyl prop-2-enoic acid, 2e-3-2-bromophenyl acrylic acid
PubChem CID 688321
IUPAC Name 3-(2-bromophenyl)prop-2-enoic acid
SMILES OC(=O)C=CC1=CC=CC=C1Br

Specifications

Melting Point 219°C
Color White-Yellow
Quantity 5 g
Formula Weight 227.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromocinnamic Acid