missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobutyryl Chloride 98.0+%, TCI America™

Catalog No. p-7123353
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41055G 5 g
B410525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41055G Supplier TCI America Supplier No. B41055G

Chemical Identifiers

CAS 22118-12-3
Molecular Formula C4H6BrClO
Molecular Weight (g/mol) 185.445
MDL Number MFCD00055269
InChI Key DROZQELYZZXYSX-UHFFFAOYSA-N
PubChem CID 90734
IUPAC Name 2-bromobutanoyl chloride
SMILES CCC(C(=O)Cl)Br

Specifications

Color Yellow
Boiling Point 65°C
Quantity 5 g
UN Number 2920
Formula Weight 185.45
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Bromobutyryl Chloride