accessibility menu, dialog, popup

UserName

CH3BrClO

CAS RN 22118-12-3

IUPAC Name: 2-bromobutanoyl chloride
Synonyms: 2-bromo-butyryl chloride
Molecular Weight (g/mol): 185.45
Molecular Formula: C4H6BrClO
InChi Key: DROZQELYZZXYSX-UHFFFAOYNA-N
SMILES: CCC(Br)C(Cl)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results