missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromobutyryl Bromide 97.0+%, TCI America™
Supplier: TCI America B056625G
Specifications
| 2-Bromobutyryl Bromide | |
| Yellow | |
| C4H6Br2O | |
| 25 g | |
| HHKDBXNYWNUHPL-UHFFFAOYSA-N | |
| 2-bromobutanoyl bromide | |
| 117293 | |
| ≥97.0% (GC) |
| 26074-52-2 | |
| 174°C | |
| MFCD00000154 | |
| 3265 | |
| CCC(C(=O)Br)Br | |
| 229.899 | |
| 229.90 | |
| Liquid |
Chemical Identifiers
| 26074-52-2 | |
| 229.899 | |
| HHKDBXNYWNUHPL-UHFFFAOYSA-N | |
| 2-bromobutanoyl bromide |
| C4H6Br2O | |
| MFCD00000154 | |
| 117293 | |
| CCC(C(=O)Br)Br |
Safety and Handling
TSCA : No