missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobiphenyl, 98%

Catalog No. AAB2184003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2184003 1 g
AAB2184006 5 g
AAB2184014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAB2184003 Supplier Thermo Scientific Chemicals Supplier No. B2184003
Get Up to $1,800 in Chemicals with Purchase
View all promotions
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 2052-07-5 | C12H9Br | 233.11 g/mol

It is used as an intermediate of organic synthesis and pharmaceutical. It also can react with norborn-2-ene to get 5-phenyl-1,2,3,4,4a,8b-hexahydro-1,4-methanobiphenylene, in synthesis of a novel trispirocyclic hydrocarbon. It is used as a reference compound to investigate extraction efficiency of solid-phase micro extraction fibers, based on methacrylic acid-trimethylolpropanetrimethacrylate copolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate of organic synthesis and pharmaceutical. It also can react with norborn-2-ene to get 5-phenyl-1,2,3,4,4a,8b-hexahydro-1,4-methanobiphenylene, in synthesis of a novel trispirocyclic hydrocarbon. It is used as a reference compound to investigate extraction efficiency of solid-phase micro extraction fibers, based on methacrylic acid-trimethylolpropanetrimethacrylate copolymers.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 2052-07-5
Molecular Formula C12H9Br
Molecular Weight (g/mol) 233.11
MDL Number MFCD00000065
InChI Key KTADSLDAUJLZGL-UHFFFAOYSA-N
Synonym 2-bromobiphenyl, o-bromobiphenyl, 2-bromo-biphenyl, 1,1'-biphenyl, 2-bromo, 2-bromo-1,1'-biphenyl, biphenyl, 2-bromo, 2-bromo-diphenyl, bromobiphenyl, 2-bromodiphenyl, 1,1'-biphenyl, bromo
PubChem CID 16329
IUPAC Name 1-bromo-2-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=CC=C2Br

Specifications

Melting Point 0°C to 2°C
Density 1.376
Boiling Point 298°C
Flash Point >110°C (230°F)
Refractive Index 1.629
Quantity 1 g
Beilstein 1863156
Solubility Information Insoluble in water.
Formula Weight 233.12
Percent Purity 98%
Chemical Name or Material 2-Bromobiphenyl
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-141-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only