missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzylamine, 96%

CAS: 3959-05-5 | C7H8BrN | 186.05 g/mol

$158.16 - $570.19

Chemical Identifiers

CAS 3959-05-5
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD00025572
InChI Key NOYASZMZIBFFNZ-UHFFFAOYSA-N
Synonym 2-bromobenzylamine, 2-bromophenyl methanamine, o-bromobenzylamine, 2-bromophenyl methylamine, 1-2-bromophenyl methanamine, 2-brobenzylamine, 2-bromo-benzylamine, pubchem20398, acmc-1ct06, benzenemethanamine, 2-bromo
PubChem CID 334072
IUPAC Name (2-bromophenyl)methanamine
SMILES C1=CC=C(C(=C1)CN)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1241206
View Documents
Thermo Scientific Chemicals
A1241206
5 g
Each for $158.16
Only null left
 
AAA1241214
View Documents
Thermo Scientific Chemicals
A1241214
25 g
Each for $570.19
Only null left
 
Description

Description

Pharmaceutical intermediates. As intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pharmaceutical intermediates. As intermediate in organic synthesis.

Solubility
Insoluble in Water. Soluble in Alcohol and Ether.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide..
Specifications

Chemical Identifiers

3959-05-5
186.05
NOYASZMZIBFFNZ-UHFFFAOYSA-N
334072
C1=CC=C(C(=C1)CN)Br
C7H8BrN
MFCD00025572
2-bromobenzylamine, 2-bromophenyl methanamine, o-bromobenzylamine, 2-bromophenyl methylamine, 1-2-bromophenyl methanamine, 2-brobenzylamine, 2-bromo-benzylamine, pubchem20398, acmc-1ct06, benzenemethanamine, 2-bromo
(2-bromophenyl)methanamine

Specifications

3959-05-5
Amine-like
MFCD00025572
UN2735
2-bromobenzylamine, 2-bromophenyl methanamine, o-bromobenzylamine, 2-bromophenyl methylamine, 1-2-bromophenyl methanamine, 2-brobenzylamine, 2-bromo-benzylamine, pubchem20398, acmc-1ct06, benzenemethanamine, 2-bromo
NOYASZMZIBFFNZ-UHFFFAOYSA-N
(2-bromophenyl)methanamine
334072
98%
118°C to 119°C (9 mmHg)
C7H8BrN
5 g
Air Sensitive
Insoluble in Water. Soluble in Alcohol and Ether.
C1=CC=C(C(=C1)CN)Br
186.05
186.06
2-Bromobenzylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only