missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzylamine, 96%

Catalog No. AAA1241206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1241206 5 g
AAA1241214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1241206 Supplier Thermo Scientific Chemicals Supplier No. A1241206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3959-05-5 | C7H8BrN | 186.05 g/mol

Pharmaceutical intermediates. As intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pharmaceutical intermediates. As intermediate in organic synthesis.

Solubility
Insoluble in Water. Soluble in Alcohol and Ether.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide..

Chemical Identifiers

CAS 3959-05-5
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD00025572
InChI Key NOYASZMZIBFFNZ-UHFFFAOYSA-N
Synonym 2-bromobenzylamine, 2-bromophenyl methanamine, o-bromobenzylamine, 2-bromophenyl methylamine, 1-2-bromophenyl methanamine, 2-brobenzylamine, 2-bromo-benzylamine, pubchem20398, acmc-1ct06, benzenemethanamine, 2-bromo
PubChem CID 334072
IUPAC Name (2-bromophenyl)methanamine
SMILES C1=CC=C(C(=C1)CN)Br

Specifications

Boiling Point 118°C to 119°C (9 mmHg)
Odor Amine-like
Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Insoluble in Water. Soluble in Alcohol and Ether.
Formula Weight 186.06
Percent Purity 98%
Chemical Name or Material 2-Bromobenzylamine
Hazard Category H302+H312-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only