missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzyl chloride, 97%

Catalog No. AAL1195303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1195303 1 g
AAL1195306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1195303 Supplier Thermo Scientific Chemicals Supplier No. L1195303

CAS: 578-51-8 | C7H6BrCl | 205.479 g/mol

2-Bromobenzyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.

Chemical Identifiers

CAS 578-51-8
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.479
MDL Number MFCD00040863
InChI Key DDVSFIUKWUTKES-UHFFFAOYSA-N
Synonym 1-bromo-2-chloromethyl benzene, 2-bromobenzyl chloride, o-bromobenzyl chloride, benzene,1-bromo-2-chloromethyl, benzene, 1-bromo-2-chloromethyl, o-brombenzylchlorid, acmc-1aunq, 3-bromo-2-chloromethylbenzene, 1-bromo-2-chloromethyl benzene #
PubChem CID 521816
IUPAC Name 1-bromo-2-(chloromethyl)benzene
SMILES C1=CC=C(C(=C1)CCl)Br

Specifications

Boiling Point 110°C to 111°C (15 mmHg)
Refractive Index 1.586
Quantity 1 g
UN Number UN3265
Beilstein 471701
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 205.49
Percent Purity 97%
Chemical Name or Material 2-Bromobenzyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only