missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzyl Bromide 98.0+%, TCI America™

Catalog No. p-7126761
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15145G 5 g
D151425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15145G Supplier TCI America Supplier No. D15145G

Chemical Identifiers

CAS 3433-80-5
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00000173
InChI Key LZSYGJNFCREHMD-UHFFFAOYSA-N
Synonym 2-bromobenzyl bromide, o-bromobenzyl bromide, 1-bromo-2-bromomethyl benzene, 2-bromobenzylbromide, alpha,2-dibromotoluene, benzene, 1-bromo-2-bromomethyl, alpha,o-dibromotoluene, benzene, bromo bromomethyl, alpha-2-dibromotoluene, a,o-dibromotoluene
PubChem CID 76965
IUPAC Name 1-bromo-2-(bromomethyl)benzene
SMILES C1=CC=C(C(=C1)CBr)Br

Specifications

Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 249.93
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromobenzyl Bromide