missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzyl bromide, 98%

Catalog No. p-72763
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC181790250 25 g Glass bottle
AC181791000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC181790250 Supplier Thermo Scientific Chemicals Supplier No. 181790250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3433-80-5 | C7H6Br2 | 249.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3433-80-5
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.93
MDL Number MFCD00000173
InChI Key LZSYGJNFCREHMD-UHFFFAOYSA-N
Synonym 2-bromobenzyl bromide, o-bromobenzyl bromide, 1-bromo-2-bromomethyl benzene, 2-bromobenzylbromide, alpha,2-dibromotoluene, benzene, 1-bromo-2-bromomethyl, alpha,o-dibromotoluene, benzene, bromo bromomethyl, alpha-2-dibromotoluene, a,o-dibromotoluene
PubChem CID 76965
IUPAC Name 1-bromo-2-(bromomethyl)benzene
SMILES C1=CC=C(C(=C1)CBr)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 29.0°C to 32.0°C
Color Colorless to Yellow
Boiling Point 129.0°C (19.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6170 to 1.6190
Linear Formula BrC6H4CH2Br
Quantity 25 g
Beilstein 05, 308
Formula Weight 249.93
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 2-Bromobenzyl bromide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 222-334-8
TSCA TSCA

RUO – Research Use Only