missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzyl bromide, 98%

Catalog No. AAA1128509
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1128509 10 g
AAA1128518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1128509 Supplier Thermo Scientific Chemicals Supplier No. A1128509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3433-80-5 | C7H6Br2 | 249.933 g/mol

2-Bromobenzyl bromide was used in the synthesis of substituted quinazolines and 1,2,3,4-tetrahydroquinazolines, 2- and 3-substituted indenes, tris-2-bromotribenzylamine3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzyl bromide was used in the synthesis of substituted quinazolines and 1,2,3,4-tetrahydroquinazolines, 2- and 3-substituted indenes, tris-2-bromotribenzylamine3.

Solubility
Insoluble in water.

Notes
Store away from strong bases and oxidizing agents. Incompatible with amines, alcohols, steel.

Chemical Identifiers

CAS 3433-80-5
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00000173
InChI Key LZSYGJNFCREHMD-UHFFFAOYSA-N
Synonym 2-bromobenzyl bromide, o-bromobenzyl bromide, 1-bromo-2-bromomethyl benzene, 2-bromobenzylbromide, alpha,2-dibromotoluene, benzene, 1-bromo-2-bromomethyl, alpha,o-dibromotoluene, benzene, bromo bromomethyl, alpha-2-dibromotoluene, a,o-dibromotoluene
PubChem CID 76965
IUPAC Name 1-bromo-2-(bromomethyl)benzene
SMILES C1=CC=C(C(=C1)CBr)Br

Specifications

Melting Point 29°C to 32°C
Boiling Point 127°C to 129°C (19 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.619
Quantity 10 g
UN Number UN1759
Beilstein 971015
Solubility Information Insoluble in water.
Formula Weight 249.94
Percent Purity 98%
Chemical Name or Material 2-Bromobenzyl bromide
Show More Show Less
Hazard Category H314-H331
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 222-334-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only