missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzyl alcohol, 98%

Catalog No. p-7022787
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1570009 10 g
AAA1570018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1570009 Supplier Thermo Scientific Chemicals Supplier No. A1570009

CAS: 18982-54-2 | C7H7BrO | 187.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18982-54-2
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD00004600
InChI Key IOWGHQGLUMEZKG-UHFFFAOYSA-N
Synonym 2-bromobenzyl alcohol, 2-bromophenyl methanol, o-bromobenzyl alcohol, benzenemethanol, 2-bromo, o-bromo-benzyl alcohol, 2-bromobenzylalcohol, 2-bromophenyl methan-1-ol, o-bromobenzylalcohol, 2-bromo-benzylalcohol, pubchem3746
PubChem CID 72850
IUPAC Name (2-bromophenyl)methanol
SMILES OCC1=CC=CC=C1Br

Specifications

Melting Point 78°C to 82°C
Quantity 10 g
Beilstein 2243418
Formula Weight 187.04
Percent Purity 98%
Chemical Name or Material 2-Bromobenzyl alcohol
Hazard Category H302+H332-H319
Precautionary Statement P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
EINECSNumber 242-719-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only