missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzoyl chloride, 98%

Catalog No. AAA1452030
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1452030 250 g
AAA1452009 10 g
AAA1452018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1452030 Supplier Thermo Scientific Chemicals Supplier No. A1452030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7154-66-7 | C7H4BrClO | 219.462 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7154-66-7
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.462
MDL Number MFCD00000655
InChI Key NZCKTGCKFJDGFD-UHFFFAOYSA-N
Synonym benzoyl chloride, 2-bromo, o-bromobenzoyl chloride, benzoyl chloride, o-bromo, 2-bromo-benzoyl chloride, 2-bromobenzoylchloride, 2-bromobenzene-1-carbonyl chloride, o-brombenzoylchlorid, bromo benzoylchloride, o-bromobenzoylchloride, pubchem12510
PubChem CID 23542
IUPAC Name 2-bromobenzoyl chloride
SMILES C1=CC=C(C(=C1)C(=O)Cl)Br

Specifications

Melting Point 10°C
Density 1.674
Boiling Point 245°C
Flash Point 107°C (224°F)
Refractive Index 1.596
Quantity 250 g
UN Number UN3265
Beilstein 508506
Sensitivity Moisture sensitive
Formula Weight 219.47
Percent Purity 98%
Chemical Name or Material 2-Bromobenzoyl chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 230-507-4
RTECSNumber DM6635000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only