missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzoyl chloride, 98%

Catalog No. AC106750100
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106750100 10 mL Glass bottle
AC106750500 50 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC106750100 Supplier Thermo Scientific Chemicals Supplier No. 106750100

CAS: 7154-66-7 | C7H4BrClO | 219.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7154-66-7
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.46
MDL Number MFCD00000655
InChI Key NZCKTGCKFJDGFD-UHFFFAOYSA-N
Synonym benzoyl chloride, 2-bromo, o-bromobenzoyl chloride, benzoyl chloride, o-bromo, 2-bromo-benzoyl chloride, 2-bromobenzoylchloride, 2-bromobenzene-1-carbonyl chloride, o-brombenzoylchlorid, bromo benzoylchloride, o-bromobenzoylchloride, pubchem12510
PubChem CID 23542
IUPAC Name 2-bromobenzoyl chloride
SMILES C1=CC=C(C(=C1)C(=O)Cl)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 11.0°C
Color Colorless to Yellow
Density 1.6700g/mL
Boiling Point 245.0°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5947 to 1.5967
Linear Formula BrC6H4COCl
Quantity 10 mL
Beilstein 09, 348
Specific Gravity 1.67
Solubility Information Solubility in water: decomposes in contact with water. Other solubilities: slightly soluble in carbontetrachloride
Formula Weight 219.46
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromobenzoyl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 230-507-4
RTECSNumber DM6635000
TSCA TSCA

RUO – Research Use Only