missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzotrifluoride 98.0+%, TCI America™

Catalog No. p-7118600
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B066325G 25 g
B0663250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B066325G Supplier TCI America Supplier No. B066325G
Only null left

Chemical Identifiers

CAS 392-83-6
Molecular Formula C7H4BrF3
Molecular Weight (g/mol) 225.008
MDL Number MFCD00000373
InChI Key RWXUNIMBRXGNEP-UHFFFAOYSA-N
Synonym 2-bromobenzotrifluoride, 1-bromo-2-trifluoromethyl benzene, o-bromobenzotrifluoride, benzene, 1-bromo-2-trifluoromethyl, 2-bromo-alpha,alpha,alpha-trifluorotoluene, 2-brombenzotrifluorid, o-bromobenzyltrifluoride, o-trifluoromethyl bromobenzene, 2-bromotrifluorotoluene, 2-brombenzotrifluorid czech
PubChem CID 9806
IUPAC Name 1-bromo-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)Br

Specifications

Boiling Point 171°C
Quantity 25 g
UN Number 1993
Formula Weight 225.01
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromobenzotrifluoride
RTECSNumber XS7980000
TSCA Yes