missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzotrifluoride, 99%, Thermo Scientific Chemicals

$175.76 - $175.76

Chemical Identifiers

CAS 392-83-6
Molecular Formula C7H4BrF3
Molecular Weight (g/mol) 225.01
MDL Number MFCD00000373
InChI Key RWXUNIMBRXGNEP-UHFFFAOYSA-N
Synonym 2-bromobenzotrifluoride, 1-bromo-2-trifluoromethyl benzene, o-bromobenzotrifluoride, benzene, 1-bromo-2-trifluoromethyl, 2-bromo-alpha,alpha,alpha-trifluorotoluene, 2-brombenzotrifluorid, o-bromobenzyltrifluoride, o-trifluoromethyl bromobenzene, 2-bromotrifluorotoluene, 2-brombenzotrifluorid czech
PubChem CID 9806
IUPAC Name 1-bromo-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC106730250
SDS View Documents
Thermo Scientific Chemicals
106730250
25 g Glass bottle
Each for $175.76
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

392-83-6
225.01
RWXUNIMBRXGNEP-UHFFFAOYSA-N
9806
C1=CC=C(C(=C1)C(F)(F)F)Br
C7H4BrF3
MFCD00000373
2-bromobenzotrifluoride, 1-bromo-2-trifluoromethyl benzene, o-bromobenzotrifluoride, benzene, 1-bromo-2-trifluoromethyl, 2-bromo-alpha,alpha,alpha-trifluorotoluene, 2-brombenzotrifluorid, o-bromobenzyltrifluoride, o-trifluoromethyl bromobenzene, 2-bromotrifluorotoluene, 2-brombenzotrifluorid czech
1-bromo-2-(trifluoromethyl)benzene

Specifications

392-83-6
100.0
1.6500g/mL
51°C
98.5% min. (GC)
C7H4BrF3
BrC6H4CF3
25 g
2-bromobenzotrifluoride, 1-bromo-2-trifluoromethyl benzene, o-bromobenzotrifluoride, benzene, 1-bromo-2-trifluoromethyl, 2-bromo-alpha,alpha,alpha-trifluorotoluene, 2-brombenzotrifluorid, o-bromobenzyltrifluoride, o-trifluoromethyl bromobenzene, 2-bromotrifluorotoluene, 2-brombenzotrifluorid czech
C1=CC=C(C(=C1)C(F)(F)F)Br
225.01
225.01
Liquid
98.5
Colorless to Brown
167°C to 168°C
Authentic
Glass bottle
1.4810 to 1.483
MFCD00000373
1.65
RWXUNIMBRXGNEP-UHFFFAOYSA-N
1-bromo-2-(trifluoromethyl)benzene
9806
99%
2-Bromobenzotrifluoride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-879-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only