missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzotrifluoride, 98%

Catalog No. p-7080553
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1013414 25 g
AAA1013422 100 g
AAA1013436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1013414 Supplier Thermo Scientific Chemicals Supplier No. A1013414
Only null left

CAS: 392-83-6 | C7H4BrF3 | 225.008 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 392-83-6
Molecular Formula C7H4BrF3
Molecular Weight (g/mol) 225.008
MDL Number MFCD00000373
InChI Key RWXUNIMBRXGNEP-UHFFFAOYSA-N
Synonym 2-bromobenzotrifluoride, 1-bromo-2-trifluoromethyl benzene, o-bromobenzotrifluoride, benzene, 1-bromo-2-trifluoromethyl, 2-bromo-alpha,alpha,alpha-trifluorotoluene, 2-brombenzotrifluorid, o-bromobenzyltrifluoride, o-trifluoromethyl bromobenzene, 2-bromotrifluorotoluene, 2-brombenzotrifluorid czech
PubChem CID 9806
IUPAC Name 1-bromo-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)Br

Specifications

Density 1.662
Boiling Point 167°C to 168°C
Flash Point 68°C (154°F)
Odor Aromatic
Refractive Index 1.482
Quantity 25 g
Beilstein 1945750
Formula Weight 225.01
Percent Purity 99%
Chemical Name or Material 2-Bromobenzotrifluoride
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 206-879-9
RTECSNumber XS7980000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only