missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzonitrile 98.0+%, TCI America™

$82.79 - $204.07

Chemical Identifiers

CAS 2042-37-7
Molecular Formula C7H4BrN
Molecular Weight (g/mol) 182.02
MDL Number MFCD00001772
InChI Key AFMPMSCZPVNPEM-UHFFFAOYSA-N
Synonym o-bromobenzonitrile, 1-bromo-2-cyanobenzene, benzonitrile, 2-bromo, benzonitrile, o-bromo, 2-bromobenzenecarbonitrile, o-bromocyanobenzene, 2-cyanobromobenzene, 2-bromo-benzonitrile, bromobenzonitrile 2-, bromobenzonitrile
PubChem CID 16272
IUPAC Name 2-bromobenzonitrile
SMILES BrC1=CC=CC=C1C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B136910G
View Documents
TCI America
B136910G
10 g
Each for $82.79
Only null left
 
B136925G
View Documents
TCI America
B136925G
25 g
Each for $204.07
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2042-37-7
182.02
AFMPMSCZPVNPEM-UHFFFAOYSA-N
16272
BrC1=CC=CC=C1C#N
C7H4BrN
MFCD00001772
o-bromobenzonitrile, 1-bromo-2-cyanobenzene, benzonitrile, 2-bromo, benzonitrile, o-bromo, 2-bromobenzenecarbonitrile, o-bromocyanobenzene, 2-cyanobromobenzene, 2-bromo-benzonitrile, bromobenzonitrile 2-, bromobenzonitrile
2-bromobenzonitrile

Specifications

2042-37-7
White-Yellow
C7H4BrN
10 g
AFMPMSCZPVNPEM-UHFFFAOYSA-N
2-bromobenzonitrile
16272
≥98.0% (GC)
2-Bromobenzonitrile
54°C
253°C
MFCD00001772
o-bromobenzonitrile, 1-bromo-2-cyanobenzene, benzonitrile, 2-bromo, benzonitrile, o-bromo, 2-bromobenzenecarbonitrile, o-bromocyanobenzene, 2-cyanobromobenzene, 2-bromo-benzonitrile, bromobenzonitrile 2-, bromobenzonitrile
BrC1=CC=CC=C1C#N
182.02
182.02
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4294

RTECSNumber : DL2459750

TSCA : No

SDS
Documents

Documents