missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzonitrile, 99%

Catalog No. AC106690100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106690100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC106690100 Supplier Thermo Scientific Chemicals Supplier No. 106690100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2042-37-7 | C7H4BrN | 182.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2042-37-7
Molecular Formula C7H4BrN
Molecular Weight (g/mol) 182.02
MDL Number MFCD00001772
InChI Key AFMPMSCZPVNPEM-UHFFFAOYSA-N
Synonym o-bromobenzonitrile, 1-bromo-2-cyanobenzene, benzonitrile, 2-bromo, benzonitrile, o-bromo, 2-bromobenzenecarbonitrile, o-bromocyanobenzene, 2-cyanobromobenzene, 2-bromo-benzonitrile, bromobenzonitrile 2-, bromobenzonitrile
PubChem CID 16272
IUPAC Name 2-bromobenzonitrile
SMILES BrC1=CC=CC=C1C#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 53°C to 56°C
Color White to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrC6H4CN
Quantity 10 g
Beilstein 09, 348
Formula Weight 182.02
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 2-Bromobenzonitrile, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-045-1

RUO – Research Use Only