missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzonitrile, 99%

Catalog No. AAA1067922
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1067922 100 g
AAA1067906 5 g
AAA1067914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1067922 Supplier Thermo Scientific Chemicals Supplier No. A1067922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2042-37-7 | C7H4BrN | 182.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2042-37-7
Molecular Formula C7H4BrN
Molecular Weight (g/mol) 182.02
MDL Number MFCD00001772
InChI Key AFMPMSCZPVNPEM-UHFFFAOYSA-N
Synonym o-bromobenzonitrile, 1-bromo-2-cyanobenzene, benzonitrile, 2-bromo, benzonitrile, o-bromo, 2-bromobenzenecarbonitrile, o-bromocyanobenzene, 2-cyanobromobenzene, 2-bromo-benzonitrile, bromobenzonitrile 2-, bromobenzonitrile
PubChem CID 16272
IUPAC Name 2-bromobenzonitrile
SMILES BrC1=CC=CC=C1C#N

Specifications

Melting Point 53°C to 56°C
Density 1.496
Boiling Point 251°C to 253°C
Flash Point >110°C (230°F)
Quantity 100 g
UN Number UN3439
Beilstein 2042185
Formula Weight 182.03
Percent Purity 99%
Chemical Name or Material 2-Bromobenzonitrile
Hazard Category H302+H312-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 218-045-1
RTECSNumber DI2459750
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only