missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzoic Acid 98.0+%, TCI America™

Catalog No. B055225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B055225G 25 g
B0552100G 100 g
B0552500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B055225G Supplier TCI America Supplier No. B055225G

Chemical Identifiers

CAS 88-65-3
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.019
MDL Number MFCD00002402
InChI Key XRXMNWGCKISMOH-UHFFFAOYSA-N
Synonym o-bromobenzoic acid, benzoic acid, 2-bromo, bromobenzoic acid, benzoic acid, o-bromo, 2-bromo-benzoic acid, benzoic acid, bromo, unii-az789tzs4l, az789tzs4l, ortho-bromobenzoic acid, attercop-chm at111657
PubChem CID 6940
IUPAC Name 2-bromobenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)Br

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 25 g
Formula Weight 201.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromobenzoic Acid
EINECSNumber (3)-0062
RTECSNumber DG4448035
TSCA Yes