missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzeneboronic acid, 98%

CAS: 244205-40-1 | C6H6BBrO2 | 200.83 g/mol

Supplier:  Thermo Scientific Chemicals L1858106

 View more versions of this product

Catalog No. AAL1858106

Only null left
Explore available promotions

Description

Description

2-Bromobenzeneboronic acid is used to promote greener amidations of carboxylic acids and amines in catalytic amounts. This technology avoids the requirement of preactivation of the carboxylic acid or use of coupling reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzeneboronic acid is used to promote greener amidations of carboxylic acids and amines in catalytic amounts. This technology avoids the requirement of preactivation of the carboxylic acid or use of coupling reagents.

Solubility
Soluble in methanol. Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications

Specifications

2-Bromobenzeneboronic acid
108°C to 110°C
MFCD01114672
8542615
Soluble in methanol. Slightly soluble in water.
OB(O)C1=CC=CC=C1Br
200.83
200.83
244205-40-1
C6H6BBrO2
5 g
2-bromophenyl boronic acid, o-bromophenylboronic acid, 2-bromobenzeneboronic acid, 2-bromophenylboronicacid, boronic acid, 2-bromophenyl, bromobenzeneboronic acid, pubchem13095, 2-bromo phenyl boronic acid, 2-bromo-phenyl boronic acid
PLVCYMZAEQRYHJ-UHFFFAOYSA-N
(2-bromophenyl)boronic acid
2773294
98%

Chemical Identifiers

244205-40-1
200.83
PLVCYMZAEQRYHJ-UHFFFAOYSA-N
2773294
OB(O)C1=CC=CC=C1Br
C6H6BBrO2
MFCD01114672
2-bromophenyl boronic acid, o-bromophenylboronic acid, 2-bromobenzeneboronic acid, 2-bromophenylboronicacid, boronic acid, 2-bromophenyl, bromobenzeneboronic acid, pubchem13095, 2-bromo phenyl boronic acid, 2-bromo-phenyl boronic acid
(2-bromophenyl)boronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only