missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzeneboronic acid, 98%

Catalog No. AAL1858103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1858103 1 g
AAL1858106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1858103 Supplier Thermo Scientific Chemicals Supplier No. L1858103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 244205-40-1 | C6H6BBrO2 | 200.83 g/mol

2-Bromobenzeneboronic acid is used to promote greener amidations of carboxylic acids and amines in catalytic amounts. This technology avoids the requirement of preactivation of the carboxylic acid or use of coupling reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzeneboronic acid is used to promote greener amidations of carboxylic acids and amines in catalytic amounts. This technology avoids the requirement of preactivation of the carboxylic acid or use of coupling reagents.

Solubility
Soluble in methanol. Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 244205-40-1
Molecular Formula C6H6BBrO2
Molecular Weight (g/mol) 200.83
MDL Number MFCD01114672
InChI Key PLVCYMZAEQRYHJ-UHFFFAOYSA-N
Synonym 2-bromophenyl boronic acid, o-bromophenylboronic acid, 2-bromobenzeneboronic acid, 2-bromophenylboronicacid, boronic acid, 2-bromophenyl, bromobenzeneboronic acid, pubchem13095, 2-bromo phenyl boronic acid, 2-bromo-phenyl boronic acid
PubChem CID 2773294
IUPAC Name (2-bromophenyl)boronic acid
SMILES OB(O)C1=CC=CC=C1Br

Specifications

Melting Point 108°C to 110°C
Quantity 1 g
Beilstein 8542615
Solubility Information Soluble in methanol. Slightly soluble in water.
Formula Weight 200.83
Percent Purity 98%
Chemical Name or Material 2-Bromobenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only