missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzamide, 98%

CAS: 4001-73-4 | C7H6BrNO | 200.035 g/mol

$124.62 - $444.86

Chemical Identifiers

CAS 4001-73-4
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.035
MDL Number MFCD00007969
InChI Key NHNAEZDWNCRWRW-UHFFFAOYSA-N
Synonym benzamide, 2-bromo, o-bromobenzamide, benzamide, o-bromo, 2-bromo-benzamide, 2-bromobenzamide, bromobenzamide, pubchem3733, maybridge1_005867, acmc-209j9x
PubChem CID 77616
IUPAC Name 2-bromobenzamide
SMILES C1=CC=C(C(=C1)C(=O)N)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1833106
View Documents
Thermo Scientific Chemicals
A1833106
5 g
Each for $124.62
Only null left
 
AAA1833114
View Documents
Thermo Scientific Chemicals
A1833114
25 g
Each for $444.86
Only null left
 
Description

Description

2-Bromobenzamide has been used in microwave assisted one-pot synthesis of substituted 3-(phenylmethylene)isoindolin-1-ones, palladium-catalyzed synthesis of phenanthridinones and in the synthesis of new water-soluble iminophosphorane ligand.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzamide has been used in microwave assisted one-pot synthesis of substituted 3-(phenylmethylene)isoindolin-1-ones, palladium-catalyzed synthesis of phenanthridinones and in the synthesis of new water-soluble iminophosphorane ligand.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

4001-73-4
200.035
NHNAEZDWNCRWRW-UHFFFAOYSA-N
77616
C1=CC=C(C(=C1)C(=O)N)Br
C7H6BrNO
MFCD00007969
benzamide, 2-bromo, o-bromobenzamide, benzamide, o-bromo, 2-bromo-benzamide, 2-bromobenzamide, bromobenzamide, pubchem3733, maybridge1_005867, acmc-209j9x
2-bromobenzamide

Specifications

4001-73-4
C7H6BrNO
5 g
Slightly soluble in water.
C1=CC=C(C(=C1)C(=O)N)Br
200.035
200.03
2-Bromobenzamide
160°C to 162°C
MFCD00007969
benzamide, 2-bromo, o-bromobenzamide, benzamide, o-bromo, 2-bromo-benzamide, 2-bromobenzamide, bromobenzamide, pubchem3733, maybridge1_005867, acmc-209j9x
NHNAEZDWNCRWRW-UHFFFAOYSA-N
2-bromobenzamide
77616
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 223-650-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only