missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzamide, 98%

Catalog No. AAA1833114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1833114 25 g
AAA1833106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1833114 Supplier Thermo Scientific Chemicals Supplier No. A1833114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4001-73-4 | C7H6BrNO | 200.035 g/mol

2-Bromobenzamide has been used in microwave assisted one-pot synthesis of substituted 3-(phenylmethylene)isoindolin-1-ones, palladium-catalyzed synthesis of phenanthridinones and in the synthesis of new water-soluble iminophosphorane ligand.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzamide has been used in microwave assisted one-pot synthesis of substituted 3-(phenylmethylene)isoindolin-1-ones, palladium-catalyzed synthesis of phenanthridinones and in the synthesis of new water-soluble iminophosphorane ligand.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 4001-73-4
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.035
MDL Number MFCD00007969
InChI Key NHNAEZDWNCRWRW-UHFFFAOYSA-N
Synonym benzamide, 2-bromo, o-bromobenzamide, benzamide, o-bromo, 2-bromo-benzamide, 2-bromobenzamide, bromobenzamide, pubchem3733, maybridge1_005867, acmc-209j9x
PubChem CID 77616
IUPAC Name 2-bromobenzamide
SMILES C1=CC=C(C(=C1)C(=O)N)Br

Specifications

Melting Point 160°C to 162°C
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 200.03
Percent Purity 98%
Chemical Name or Material 2-Bromobenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-650-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only