missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromobenzaldehyde 98.0+%, TCI America™
Supplier: TCI America B083625G
Specifications
| 2-Bromobenzaldehyde | |
| Yellow | |
| C7H5BrO | |
| 25 g | |
| NDOPHXWIAZIXPR-UHFFFAOYSA-N | |
| 2-bromobenzaldehyde | |
| 81129 | |
| ≥98.0% (GC) |
| 6630-33-7 | |
| 230°C | |
| MFCD00003300 | |
| o-bromobenzaldehyde, benzaldehyde, 2-bromo, benzaldehyde, o-bromo, 2-bromo benzaldehyde, bromobenzaldehyde, bromo benzaldehyde, o-bromo-benzaldehyde, 2-formylbromobenzene, 2-bromo-benzaldehyde, benzaldehyde, bromo | |
| BrC1=CC=CC=C1C=O | |
| 185.02 | |
| 185.02 | |
| Liquid |
Chemical Identifiers
| 6630-33-7 | |
| 185.02 | |
| NDOPHXWIAZIXPR-UHFFFAOYSA-N | |
| 81129 | |
| BrC1=CC=CC=C1C=O |
| C7H5BrO | |
| MFCD00003300 | |
| o-bromobenzaldehyde, benzaldehyde, 2-bromo, benzaldehyde, o-bromo, 2-bromo benzaldehyde, bromobenzaldehyde, bromo benzaldehyde, o-bromo-benzaldehyde, 2-formylbromobenzene, 2-bromo-benzaldehyde, benzaldehyde, bromo | |
| 2-bromobenzaldehyde |
Safety and Handling
TSCA : Yes