missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromoanthraquinone 96.0+%, TCI America™
$77.00 - $1409.23
Chemical Identifiers
| CAS | 572-83-8 |
|---|---|
| Molecular Formula | C14H7BrO2 |
| Molecular Weight (g/mol) | 287.112 |
| InChI Key | VTSDGYDTWADUJQ-UHFFFAOYSA-N |
| PubChem CID | 221013 |
| IUPAC Name | 2-bromoanthracene-9,10-dione |
| SMILES | C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)Br |
Chemical Identifiers
| 572-83-8 | |
| 287.112 | |
| 221013 | |
| C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)Br |
| C14H7BrO2 | |
| VTSDGYDTWADUJQ-UHFFFAOYSA-N | |
| 2-bromoanthracene-9,10-dione |
Specifications
| 572-83-8 | |
| C14H7BrO2 | |
| VTSDGYDTWADUJQ-UHFFFAOYSA-N | |
| 2-bromoanthracene-9,10-dione | |
| 221013 | |
| ≥96.0% (GC) | |
| 2-Bromoanthraquinone |
| 208°C | |
| 1 g | |
| C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)Br | |
| 287.112 | |
| 287.11 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : CB5950000
TSCA : No