missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoanisole 98.0+%, TCI America™

Catalog No. p-7118513
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B054625G 25 g
B0546100G 100 g
B0546500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B054625G Supplier TCI America Supplier No. B054625G
Only null left

Chemical Identifiers

CAS 578-57-4
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD00000064
InChI Key HTDQSWDEWGSAMN-UHFFFAOYSA-N
Synonym 2-bromoanisole, o-bromoanisole, benzene, 1-bromo-2-methoxy, 2-methoxybromobenzene, o-anisyl bromide, anisole, o-bromo, o-methoxybromobenzene, 2-bromoanisol, o-methoxyphenyl bromide, 2-methoxyphenyl bromide
PubChem CID 11358
IUPAC Name 1-bromo-2-methoxybenzene
SMILES COC1=CC=CC=C1Br

Specifications

Color Yellow
Boiling Point 223°C
Quantity 25 g
Formula Weight 187.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromoanisole
RTECSNumber BZ8500000
TSCA Yes