missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromoanisole, 98%
Description
2-Bromoanisole was used in the synthesis of unsymmetrical substituted biphenyl compounds, used to produce (2-methoxy-phenyl)-acetone at the temperature of 100°C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromoanisole was used in the synthesis of unsymmetrical substituted biphenyl compounds, used to produce (2-methoxy-phenyl)-acetone at the temperature of 100°C.
Solubility
Soluble in ethanol and diethyl ether.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
2-Bromoanisole was used in the synthesis of unsymmetrical substituted biphenyl compounds, used to produce (2-methoxy-phenyl)-acetone at the temperature of 100°C.
Solubility
Soluble in ethanol and diethyl ether.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Specifications
| Melting Point | 2°C to 3°C |
| Density | 1.511 |
| Boiling Point | 220°C to 222°C |
| Flash Point | 96°C (204°F) |
| Assay Percent Range | 98% |
| Refractive Index | 1.574 |
| Quantity | 100 g |
| Beilstein | 1859996 |
| Solubility Information | Soluble in ethanol and diethyl ether. |
| Formula Weight | 187.04 |
| Show More |
Safety and Handling
| EINECSNumber | 209-425-8 |
| RTECSNumber | BZ8500000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only