missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoanisole, 98%

Catalog No. AAA1169214
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1169214 25 g
AAA1169222 100 g
AAA1169236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1169214 Supplier Thermo Scientific Chemicals Supplier No. A1169214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 578-57-4 | C7H7BrO | 187.036 g/mol

2-Bromoanisole was used in the synthesis of unsymmetrical substituted biphenyl compounds, used to produce (2-methoxy-phenyl)-acetone at the temperature of 100°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoanisole was used in the synthesis of unsymmetrical substituted biphenyl compounds, used to produce (2-methoxy-phenyl)-acetone at the temperature of 100°C.

Solubility
Soluble in ethanol and diethyl ether.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 578-57-4
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD00000064
InChI Key HTDQSWDEWGSAMN-UHFFFAOYSA-N
Synonym 2-bromoanisole, o-bromoanisole, benzene, 1-bromo-2-methoxy, 2-methoxybromobenzene, o-anisyl bromide, anisole, o-bromo, o-methoxybromobenzene, 2-bromoanisol, o-methoxyphenyl bromide, 2-methoxyphenyl bromide
PubChem CID 11358
IUPAC Name 1-bromo-2-methoxybenzene
SMILES COC1=CC=CC=C1Br

Specifications

Melting Point 2°C to 3°C
Density 1.511
Boiling Point 220°C to 222°C
Flash Point 96°C (204°F)
Assay Percent Range 98%
Refractive Index 1.574
Quantity 25 g
Beilstein 1859996
Solubility Information Soluble in ethanol and diethyl ether.
Formula Weight 187.04
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromoanisole
Show More Show Less
EINECSNumber 209-425-8
RTECSNumber BZ8500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only