missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromoacetyl)thiophene, 97%, Thermo Scientific Chemicals

Catalog No. AAH5106006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5106006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5106006 Supplier Thermo Scientific Chemicals Supplier No. H5106006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10531-41-6 | Molecular Formula: C6H5BrOS | Molecular Weight: 205.069 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10531-41-6
Molecular Formula C6H5BrOS
Molecular Weight (g/mol) 205.069
MDL Number MFCD02677721
InChI Key UHWNENCHFSDZQP-UHFFFAOYSA-N
Synonym 2-bromo-1-thiophen-2-yl ethanone, 2-2-bromoacetyl thiophene, 2-bromo-1-2-thienyl-1-ethanone, 2-bromoacetyl thiophene, 2-bromo-1-thiophen-2-yl-ethanone, 2-bromo-1-2-thienyl ethanone, 2-bromo-1-thiophen-2-yl ethan-1-one, ethanone, 2-bromo-1-2-thienyl, ethanone, 2-bromo-1-thienyl, 2-bromo-1-2-thienyl ethan-1-one
PubChem CID 2776372
IUPAC Name 2-bromo-1-thiophen-2-ylethanone
SMILES C1=CSC(=C1)C(=O)CBr

Specifications

Melting Point 29°C to 33°C
Density 1.62
Boiling Point 127°C to 130°C (7 mmHg)
Odor Pungent
Quantity 5 g
UN Number UN3261
Sensitivity Air and moisture sensitive
Formula Weight 205.07
Percent Purity 97%
Chemical Name or Material 2-(Bromoacetyl)thiophene
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only