accessibility menu, dialog, popup

UserName

SOBr

CAS RN 10531-41-6

IUPAC Name: 2-bromo-1-(thiophen-2-yl)ethan-1-one
Synonyms: 2-bromo-1-(thiophen-2-yl)ethanone
Molecular Weight (g/mol): 205.07
Molecular Formula: C6H5BrOS
InChi Key: UHWNENCHFSDZQP-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=CS1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results