missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromoacetyl)-6-methoxynaphthalene 98.0+%, TCI America™

Catalog No. B34961G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34961G 1 g
B34965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34961G Supplier TCI America Supplier No. B34961G

Chemical Identifiers

CAS 10262-65-4
Molecular Formula C13H11BrO2
Molecular Weight (g/mol) 279.133
MDL Number MFCD03964189
InChI Key HHHKEQGAGUAOQI-UHFFFAOYSA-N
Synonym 2-bromoacetyl-6-methoxynaphthalene, 2-bromo-1-6-methoxy-2-naphthyl ethanone, 2-bromo-6'-methoxy-2'-acetonaphthone, 2-bromo-1-6-methoxynaphthalen-2-yl ethan-1-one, 2-bromo-1-6-methoxynaphthalen-2-yl ethanone, ethanone, 2-bromo-1-6-methoxy-2-naphthalenyl, enamine_005243, 2-bromo-1-6-methoxy-naphthalen-2-yl-ethanone
PubChem CID 193452
IUPAC Name 2-bromo-1-(6-methoxynaphthalen-2-yl)ethanone
SMILES COC1=CC2=C(C=C1)C=C(C=C2)C(=O)CBr

Specifications

Melting Point 112°C
Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 279.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Bromoacetyl)-6-methoxynaphthalene